Safety pipette filler, Flip
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4713611 4713641 4713621
612-1945EA 8.8 EUR
612-1945 612-1948 DENE4713621
Safety pipette filler, Flip
Pipetten Pipetvullerballonnen
Natural rubber.
  • Simple, one-handed design
  • Only two operating points
  • Easy to evacuate via automatic valve
  • Removable valve for easy inside cleaning and rinsing