Maatcilinders, lage vorm, klasse B, borosilicaatglas
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
432341544 632432341123 632432341338 632432341125 432341443
612-3800EA 88.2 EUR
612-3800 612-3778 612-3798 612-3779 612-3799
Maatcilinders, lage vorm, klasse B, borosilicaatglas
Cilinders Maatcilinders
Borosilicate glass, low form, class B.
  • Hexagonal base
  • White graduations
Certificaten: ISO 4788, DIN 12 680-1