Bulb pipettes, works certified, MBL, one mark, class AS
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PRA742ASWC PRA740ASWC PRA744ASWC PRA746ASWC PRA748ASWC PRA750ASWC PRA752ASWC PRA754ASWC PRA756ASWC
612-3898EA 18.2 EUR
612-3898 612-3897 612-3899 612-3908 612-3909 612-3910 612-3911 612-3924 612-3925
Bulb pipettes, works certified, MBL, one mark, class AS
Pipetten Volpipetten
Soda lime glass.
  • Each pipette engraved with individual serial number
  • Markings in permanent amber stain
Certificaten: ISO 648, DIN 12690
Leveringsinformatie: Supplied with calibration certificate allowing traceability to National Standards.