Maatkolven, klasse B
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
67695
612-3916EA 28.2 EUR
612-3916
Maatkolven, klasse B
Kolven Maatkolven
PMP, translucent, with PP NS-stopper.
  • Tolerances are approximately 20% better than class B according to DIN EN ISO 1042
  • Individually ring marked
  • With NS stoppers
Certificaten: DIN EN ISO 1042