Bulb pipette, MBL®, (one mark), class B
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PRA700B PRA702B PRA704B PRA706B PRA708B PRA712B PRA714B PRA716B PRA710B
612-3926EA 19.3 EUR
612-3926 612-3927 612-3928 612-3929 612-3930 612-3937 612-3938 612-3939 612-3936
Bulb pipette, MBL®, (one mark), class B
Pipetten Volpipetten
Soda-lime glass.
  • Calibrated to deliver (TD)
  • Markings in permanent amber stain
Certificaten: ISO 648.