Graduated pipettes, type 1, Class B, MBL
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PRA760B PRA762B PRA764B PRA766B PRA768B
612-3950EA 20 EUR
612-3950 612-3951 612-3952 612-3953 612-3954
Graduated pipettes, type 1, Class B, MBL
Pipetten Gegradueerde pipetten
Soda lime glass.
  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain
DIN EN ISO 835
Certificaten: DIN EN ISO 835.