Measuring cylinder, tall form, class B, PP
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
646941 647941 648941 649941 650941 651941 652941 653941
612-2426EA 3 EUR
612-2426 612-2427 612-2428 612-2429 612-4354 612-4355 612-4356 612-4357
Measuring cylinder, tall form, class B, PP
Cilinders Maatcilinders
Doorschijnend PP, hoog model, klasse B.
  • Calibrated to 'In'
  • Hexagonal base, ring mark on the principal points
  • Thermal stresses of up to +80 °C do not result in the tolerance limit being permanently exceeded
Certificaten: DIN 12681 B / ISO 6706
Order Now
Specificaties
Environmentally Preferable