Maatkolven, klasse A, blauwe maatverdeling
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2820166 2820197 2820198 2820152 2820153 2820160 2820161 2820165 2820170 2820171 2820175 2820178 2820180 2820181 2820185 2820186 2820190 2820192 2820193 2820194 2820182 282015208 282016008 282016108 282016508 282017008 282017508 282017808 282018008 282018108 282018508 282018608 282019008 282019308 282019408 282019445 282019708 282019808
612-5737EA 19.1 EUR
612-5737 612-3307 612-3308 612-1556 612-1557 612-1558 612-1559 612-1562 612-1563 612-1564 612-1565 612-1566 612-1567 612-1568 612-1569 612-1570 612-1571 612-1572 612-1573 612-1574 612-5872 612-5863 612-5864 612-5865 612-5866 612-5867 612-5868 612-5869 612-5870 612-5871 612-5873 612-5874 612-5875 612-5876 612-5877 612-5878 612-5879 612-5880
Maatkolven, klasse A, blauwe maatverdeling
Kolven Maatkolven
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Blue enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website (www.hirschmannlab.de)
* In aanvulling op DIN.
**Brede hals
Certificaten: DIN EN ISO 1042