VWR® Inoculating Loops and Needles, Disposable
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-7259EA 30.5 EUR
612-7259 612-7260 612-7262 612-7263 612-7264 612-7265 612-7266 612-7268 612-7270 612-7271 612-7272 612-7273 612-7274 612-7277 612-7278
VWR® Inoculating Loops and Needles, Disposable
Uitstrijkspatels Entogen en -naalden
Inoculating loops and needles are devices used for inoculation, serial dilution, sterile sampling, transfer, and microbiological sample spreading.
  • PS, sterile
  • Colour coded for easy product identification
  • Hexagonal loop shaft improves grip, assisting orientation and making diluting and streaking easier
  • E-beam sterilised
The resealable pouch allows loops to be easily accessed any number of times, then re-closed to maintain product integrity. The loops and needles are positioned upside down in the pouch in order to reduce any risk of contamination.More Product Information