Burettes, lateral stopcock, Schellbach
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3210160 3210170 3210175 3210360 3210370 3210375
612-9821EA 58 EUR
612-9821 612-9822 612-9823 HIRS3210360 HIRS3210370 HIRS3210375
Burettes, lateral stopcock, Schellbach
Buretten
Burette, class B, DURAN® borosilicate glass 3.3, with blue graduation.
  • Lateral glass stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
Certificaten: Calibrated for EX, DIN EN ISO 385.