Original Perfusor® syringes
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8728844F 8728861F-06 8728852F-06 8728810F-06
613-0120EA 196 EUR
613-0120 BRAU8728861F-06 BRAU8728852F-06 BRAU8728810F-06
Original Perfusor® syringes
Spuiten Injectiespuiten, medisch
PP syringe with safe plunger backstop and minimal residual volume.
  • Oval grip plate for handling of the syringe and for prevention of rolling
  • Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material or natural latex-free
  • Centric Luer Lock cone for the attachment of a needle or infusion tubing