Elektronische meerkanaalspipetten, Transferpette® electronic
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
705403 705406 705453 705456 705400 705450 705416 705413 705410 705466 705460 705463 705409 705399 705404 705414 705449 705454 705459 705464 705398 705448 705397 705447
613-0214EA 870 EUR
613-0214 613-0215 613-0054 613-0220 613-0213 613-0219 613-0218 613-0217 613-0216 613-0223 613-0221 613-0222 613-0605 613-0604 613-0606 613-0607 613-0608 613-0610 613-0609 613-0611 613-7181 613-7164 613-7256 613-7257
Elektronische meerkanaalspipetten, Transferpette® electronic
Pipetten Micropipetten
Transferpette® electronic pipettes combines the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C)
  • Five convenient programs: Pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing
  • Easy Calibration-Technique for making quick adjustments of the Transferpette® electronic, without tools
The Transferpette® electronic is intended for the pipetting of liquids within the followingMore Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage,Duurzame verpakking