Eenkanaalspipetten, elektronisch, variabel volume, Eppendorf Xplorer®
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4861000.015 4861000.040 4861000.023 4861000.031 4861000.058 4861000.066 4861000.027 4861000.044 4861000.017
613-1398EA 722 EUR
613-1398 613-1407 613-1399 613-1400 613-1408 613-1409 613-6981 613-6982 613-6980
Eenkanaalspipetten, elektronisch, variabel volume, Eppendorf Xplorer®
Pipetten Micropipetten
Eppendorf Xplorer single-channel pipettes provide reliable, ergonomic high-precision liquid handling. With easy operation, five pipetting modes, and advanced charging technology, they minimise errors and boost reproducibility in a range of lab applications.
  • Electronic single channel pipettes designed to aspirate and dispense liquid volumes from 0,5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker with ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle and powerful rechargeable battery
  • Flexible
More Product Information