BD Discardit™ II Syringes
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
300296 300928 309040 309050 309110 300322 300512 300199 300296UK 300512UK 301360 309210
613-3916EA 30.8 EUR
613-3916 613-3932 BDAM309040 613-3951 613-3952 615-0031 615-0032 BDAM300199 BDAM300296UK BDAM300512UK BDAM301360 BDAM309210
BD Discardit™ II Syringes
Spuiten Injectiespuiten, medisch
The BD Discardit™ II two-piece syringe is a single-use medical device intended for injection and aspiration of medical fluids, including both bodily fluids (blood, etc.) and medicines. The lubricant included in the barrel material allows smooth advancement of the plunger, leading to reduced sliding force.
  • Clear and transparent barrel for good visualisation of syringe content
  • Leak-tight
  • Accurate graduation suited to user's needs
  • Robust retaining ring to prevent accidental plunger rod pullout during aspiration
  • Design of finger grip flanges ensures stability
More Product Information