VWR® Zippette Classic, Bottle-Top Dispensers
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
613-4002EA 0 EUR
613-4002
VWR® Zippette Classic, Bottle-Top Dispensers
Flesdispensers
Variable volume, autoclavable.
  • Fine adjustment for exact, reproducible dispensing
  • All wetted parts are chemically resistant and autoclavable at +121 °C
  • Easily removable PTFE piston for smooth action and easy cleaning
  • Borosilicate glass barrel protected with a transparent PP sleeve can be easily removed and cleaned
  • Easy priming and minimum waste with no leakage back into the reservoir
Accurate dispensing from an easy to use dispenser. The specially designed delivery spout ensures exact dispensing into narrow neck containers, and the anti-drip safetyMore Product Information