Charging pipette carousel for four pipettes
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
730991
613-4124EA 653 EUR
613-4124
Charging pipette carousel for four pipettes
Pipethouders
The charging carousel for four pipettes is a convenient, easy-to-use storage and charge system designed to keep your electronic pipettes ready for use and safe from contamination.
  • Charging carousel has a simple, elegant design
  • Holds pipettes in an upright position while not in use to avoid contamination from benches and tables
  • Compact carousel stand: Saves valuable bench space
Proper use and storage will ensure long-lasting use of your electronic pipettes, keeping them ready for whenever you need them. When storing your electronic pipette (or anyMore Product Information
Order Now
Specificaties
Environmentally Preferable