Volumetric flasks, class A, brown markings
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3.664.016.A
613-4125EA 21.2 EUR
613-4125
Volumetric flasks, class A, brown markings
Kolven Maatkolven
DURAN® borosilicate glass 3.3 with PE stopper.
  • With spout
  • Conformity certified
  • Delivered with batch number and have enclosed one batch certificate per package