Dispensers, Acurex™ 501, Socorex

Leverancier: Socorex Isba
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

501.02025 501.021 501.022 501.0505 501.051 501.052 501.101 501.102 501.302
613-4837EA 439 EUR
613-4837 613-4838 613-4839 613-4840 613-4841 613-4842 613-4843 613-4844 613-4845
Dispensers, Acurex™ 501, Socorex
Flesdispensers
Variabel volume, autoclaveerbaar.

  • Dosing mechanism protected inside reservoir
  • Retractable graduated column reducing instrument height
  • Fluid path made of inert materials excluding any metal
  • Amber glass adequate for routine use with optimal light protection
  • Choice of four reservoir sizes: 250, 500, 1000 and 2000 ml

Low profile dispensers for safe reagent handling adapted to refrigerator storage as well as water bath heating.

Volume: 0,2 to 30 ml

Leveringsinformatie: Delivered with a QC certificate (lot identification on the instruments).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR