Single channel macro pipettes, fixed / variable volume, Acura® manual 835F and 835
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
835.F10 835.F05 835.F02.5 835.05 835.10 835.F02 835.02 835.05PP 835.02PP
SOCO835.F10EA 297 EUR
SOCO835.F10 SOCO835.F05 SOCO835.F02.5 613-4879 613-4880 SOCO835.F02 SOCO835.02 SOCO835.05PP SOCO835.02PP
Single channel macro pipettes, fixed / variable volume, Acura® manual 835F and 835
Pipetten Micropipetten
These mechanical pipettes are precision instruments combining high-tech materials with user friendly ergonomics.
  • Interchangeable nozzle filters for added protection against liquid overflow and barrel contamination
  • Optional adapters allow fitting of glass pasteur pipettes on 2 and 5 ml models in addition to PP tips
  • Precision digital display visible at all time
  • Adjustable, soft padded tip ejector (Justip™) fitting most tips
  • Shock, UV-light and autoclaving resistance
  • Tight lip seal and new springs provide for ultra-soft pipetting
  • Autoclavable
Models withMore Product Information
Order Now
Specificaties
Environmentally Preferable