tempmate® M2T Multi-Use Temperature Data Logger
Leverancier: tempmate
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M2T-000
620-2826EA 46.8 EUR
620-2826
tempmate® M2T Multi-Use Temperature Data Logger
Dataloggers
Intuitive multi-use temperature logger for reliable low-cost cold chain monitoring. For temperature and humidity recording with internal or external temperature and humidity sensor, multifunction display and automatic PDF report generation.
  • Product-related temperature monitoring along the entire cold chain, in warehouses or fridges
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • Free available software for individual logger configuration
  • Detailed PDF report including graph and table
  • Multifunction display with
More Product Information