Benzoic acid

Leverancier: Mettler - Toledo
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

00018555
621-0032EA 0 EUR
621-0032
Benzoic acid
Benzoic acid
Formule: C₇H₆O₂
Kookpunt: 249 °C (1013 hPa)
Smeltpunt: 121…123 °C
Dichtheid: 1,316 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002398
CAS nummer: 65-85-0
EINECS: 200-618-2
Bereik: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR