Stereo microscopes, SMZ160 series
Leverancier: MOTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1100201300151 1100201300171 1100201300161 1100201300181 1101001403911 1101001403931 1101001403941 1101001708251 1101001708261 1101001708271 1101001708281 1101001708291 1101010100911 1101010100921
630-3493EA 639 EUR
630-3493 630-3495 630-3494 630-3496 630-3497 630-3498 630-3499 630-3506 630-3507 630-3508 630-3509 630-3510 630-3511 630-3512
Stereo microscopes, SMZ160 series
Microscopen
A versatile stereomicroscope for everyday use in education, industry, and the life sciences. Model SMZ160 features Greenough optical system, 6:1 Zoom ratio, 0.75x / 4.5X Zoom magnification, working distance 100mm, a pole type stand with LED incident and transmitted light or no light integrated. Expand your possibilities with a wide range of accessories.
  • Magnification: Continous zoom from 7,5× - 45×
  • Binocular or trinocular head to mount a digital camera
  • Available with LED illuminated or plain stands without light
  • Implemented click-stops allow precise
More Product Information