Staining racks
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
41185010 41186010 41187010
631-9156EA 96.4 EUR
631-9156 631-0336 631-0337
Staining racks
Rekken Kleurrekken
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.