Staining systems for microscope slides, DrainRack Junior
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M710-50W M710-50P M710-50Y M710-50B M710-50G
631-1918EA 59.6 EUR
631-1918 631-1917 631-1919 631-1915 631-1916
Staining systems for microscope slides, DrainRack Junior
Kleurbakken
This rugged tray used as a drain rack can hold up to 100 microscope slides in 50 individually numbered slots. All slides are stored upright for easier insertion and removal.
  • Barcodes can be read without having to remove the slides from the box
  • To save space, the number of slides in each slot can be doubled simply by inserting two slides in each slot
  • Not autoclavable
Order Now
Specificaties
Environmentally Preferable