Platinum wire
Leverancier: CHEMPUR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PT02 900427 PT06 009348 009349
631-7101EA 0 EUR
631-7101 631-7102 631-7105 631-7106 631-7107
Platinum wire
Draad
Platinum 99,9%.
Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.