NMR tubes, high-throughput, thin walled, 5 mm Ø, SP Wilmad-LabGlass

Leverancier: SP Industries
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

WG-1000-7 WG-1000-8 WG-1000-4
634-0871EA 180 EUR
634-0871 634-0872 634-1157
NMR tubes, high-throughput, thin walled, 5 mm Ø, SP Wilmad-LabGlass
Buisjes NMR-buisjes
Borosilicate glass, class B, ASTM E438 Type 1

  • Best Ø tolerance in the industry and a 30% thicker wall than any competitors' product
  • Manufactured in an ISO 9001:2008 USA facility
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • With or without disposable cap

Thin walled high throughput NMR tubes have an average camber of 60 µm to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR