Spectrophotometer cuvettes, NanoCuvette™ One
Leverancier: cphnano
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17 3
634-1176EA 0 EUR
634-1176 634-1177
Spectrophotometer cuvettes, NanoCuvette™ One
Cuvetten Cuvetten voor spectrofotometers
Get accurate quantification by upgrading a UV-Vis spectrophotometer with user-friendly cuvettes. The NanoCuvette™ One expands the capabilities of an UV/Vis spectrophotometer to include quantification via refractive index for QC, kinetics, and 0,5 μl sample measurements.
  • Excellent for QC, kinetics, and label-free concentrations of a wide range of samples
  • All major UV/Vis spectrophotometer brands supported seamlessly out of the box
  • Based on semi-disposable PMMA cuvette, and reusable for low-cost measurements
  • Results independent of cleaning, pipetting errors,
More Product Information