Digital pocket refractometers for pharmaceutical ingredients, PAL series
Leverancier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4425 4426 4431 4432 4451 4455 4464 4465 4466 4468 4470 4471 4472 4413
635-0232EA 308 EUR
635-0232 635-0233 635-0237 635-0238 635-0255 635-0259 635-0268 635-0269 635-0270 635-0272 635-0274 635-0275 635-0276 635-0221
Digital pocket refractometers for pharmaceutical ingredients, PAL series
refractometers Draagbare refractometers
The PAL series for pharmaceutical ingredients (reagents) are pocket instruments delivering rapid results.
Certificaten: IP 65 allows rinsing off the sample with running water.