Handheld refractometers for wine
Leverancier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4484 4479 4480 4483 4486 4487
635-0157EA 308 EUR
635-0157 635-0277 635-0278 635-0279 635-0281 635-0282
Handheld refractometers for wine
refractometers Draagbare refractometers
The PAL series with special scales are pocket sized.
  • Simple handed operation
  • Rapid results: Three second response
  • Battery operated instruments
Certificaten: IP 65 allows rinsing off the sample with running water.