Immersion flow cups, ISO 2431
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2093 VF2092 VF2091 VF2090 VF2224 VF2222 VF2185
649-0010EA 177 EUR
649-0010 649-0009 649-0008 649-0007 649-0014 649-0013 649-0011
Immersion flow cups, ISO 2431
Viscosimeters Flow-cup viscometers
Titanium anodized aluminum or stainless steel cups designed with fixed stainless steel nozzle and handle, for measuring coatings and other fluids.
Certificaten: ISO 2431