Titrator, Karl Fischer, volumetrisch, TitroLine® 7800
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285220980 285220990 285221030
661-0239EA 6220 EUR
661-0239 661-0240 661-0242
Titrator, Karl Fischer, volumetrisch, TitroLine® 7800
Titratieapparaten
The Titroline® 7800 enhanced the universal features of the TitroLine® 7750 (potentiometric titration and volumetric KF titration), with an additional IDS measurement socket. Hence the TitroLine® 7800 is able to perform potentiometric titrations with analogue or IDS® electrodes as well as volumetric Karl Fischer titrations. The digital (IDS) measurement socket allows the connection of various digital sensors, such as for measuring of conductivity.
  • Twee meetingangen: 1× analoog en 1× digitaal voor intelligente digitale sensoren (IDS®)
  • IDS®-sensoren bieden de
More Product Information