pH-buffers, klaar voor gebruik

Leverancier: WTW
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

108706 108708 108800 108802 108805 108703 108698 108700 108702 108704 108722 108825 108811 108812 108824 108827 108801 108803 108809 108814 108826
662-0319EA 15.1 EUR
662-0319 662-0320 662-8222 662-8224 WTWA108805 662-0270 662-0329 662-8232 662-8234 662-8236 WTWA108722 WTWA108825 WTWA108811 WTWA108812 WTWA108824 WTWA108827 WTWA108801 WTWA108803 WTWA108809 WTWA108814 WTWA108826
pH-buffers, klaar voor gebruik
Buffers pH-buffers

Every bottle has its own lot reference with the expiration date.


  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample


Accuracy is ±0,03 pH


Certificaten: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR