Sodium calibration standards for ISE measurement, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
941105 941107 650700 841108 841109
662-0831EA 156 EUR
662-0831 662-0832 ORIO650700 662-0744 ORIO841109
Sodium calibration standards for ISE measurement, Orion™
Standaarden Kationische standaarden Natrium standaarden
Solutions for 8611BNWP Combination or 8411BN & 800300 Half-cell Electrodes.