Micro pH elektroden, Orion
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9810BN
662-0886EA 1050 EUR
662-0886
Micro pH elektroden, Orion
Elektroden pH-elektroden
pH electrodes with BNC connector and 1 m cable.
  • pH range: 0 to 14
  • Material: Glass
Order Now
Specificaties
Environmentally Preferable