Buffer solutions in sachets, perpHect® Pouches, Orion™

Leverancier: Thermo Orion
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

910725 910410-WA 910425 910710 911010
662-0780EA 94.9 EUR
662-0780 662-1143 662-0777 662-1144 662-1147
Buffer solutions in sachets, perpHect® Pouches, Orion™
Buffers pH-buffers

Solutions in sachets ensure optimum precision. The sachet provides a perfect liquid seal and good protection from light.


  • Disposable
  • Convenient, colour coded sachet and solution for ease of use
  • No danger of contamination
  • Traceable to NIST standards


Colour coded: pH 4,01 buffer is pink, pH 7,00 buffer is yellow, and pH 10,01 buffer is blue.


There is no wasted buffer or secondary reservoir. Each sachet contains lot code, expiration date, and buffer temperature table.


The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR