VWR®, pHenomenal® pH-elektroden
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
620-1659EA 186 EUR
620-1659 662-1162 662-1163 662-1164 662-1158 662-1159 662-1161 662-1156 662-1157 662-1247
VWR®, pHenomenal® pH-elektroden
Elektroden pH-elektroden
pH-elektroden met BNC-connector en kabel.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Leveringsinformatie: Inclusief alleen pH-elektrode. 620-1659 bevat alleen pHenomenal® TEMP21 T-sensor (NTC 30 kOhm).
Order Now
Specificaties
Environmentally Preferable