Gecombineerde pH-elektroden
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51109503 51109505 51109507 51109502 51109500 51109504 51109502
662-2161EA 708 EUR
662-2161 662-2163 662-1894 662-2160 662-2159 662-1669 662-2478
Gecombineerde pH-elektroden
Elektroden pH-elektroden
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
  • Integrated glass sleeve for secure fixation in the titration head
  • With S7 screw cap and detachable cap or with Plug & Play S12 connection
  • Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111More Product Information