Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Leverancier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ELITEPH ELITEPHKIT ELITEPHSPEAR ELITEPHSPEARKI
662-2303EA 148 EUR
662-2303 662-2280 662-2304 662-2282
Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Elektrochemische meters pH- en multiparametersystemen
These rugged and user-friendly testers feature a simultaneous display of pH and temperature and provide accurate measurements for water analysis. Available in a standard version or with a spear tip open pore sensor for direct measurements of solid or semi-solid samples such as cheese, meat, fruit, bread and more.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; one-push calibration
  • Easy to replace sensor
  • Lanyard provision for ‘on the go’ measurements
  • IP 67 waterproof rating
  • Standard version only: Leakproof cap with fill-line
More Product Information