Bioproces steriliseerbare pH- en ORP-elektroden, SteamLine
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285113213 285113246 285113254 285113262 285113402 285113419 285113427 285113443 285113468 285113398 285113381 285113373 285113365 285113316 285113308 285114025 285114035 285114065 285114075 285114085 285113332 285114045 285113324 285114055 285113400 285113390 285113585
662-6145EA 350 EUR
662-6145 662-6146 662-6147 662-6149 662-6159 662-6160 662-6161 662-6162 662-6148 SCOI285113398 SCOI285113381 SCOI285113373 SCOI285113365 662-1966 662-1833 662-2277 SCOI285114035 SCOI285114065 SCOI285114075 SCOI285114085 662-2315 SCOI285114045 662-2314 SCOI285114055 SCOI285113400 SCOI285113390 SCOI285113585
Bioproces steriliseerbare pH- en ORP-elektroden, SteamLine
Elektroden pH-elektroden
Autoclavable high-tech electrodes with outstanding electrical and physical performance characteristics guaranteeing exceptional accuracy and reliability over long periods of time. Can be used in demanding environments such as SIP treatment (Sterilisation in Place), and CIP cleaning (Clean in Place).
  • Low maintenance electrodes
  • Pressurised reference electrode allowing a significant longer operational time of the electrode
  • S-membrane glass avoiding damages from hot caustic soda
  • Allows up to 100 sterilisations with overheated water vapour at about 135 °C
  • For
More Product Information