Conductivity meter, handheld, Eutech COND 6+
Leverancier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ECCON603PLUS ECCON603PLUSK
663-0350EA 478 EUR
663-0350 663-0353
Conductivity meter, handheld, Eutech COND 6+
Elektrochemische meters Geleidbaarheidsmeters
This portable economic meter with large LCD display is ideal for quick, accurate conductivity checks in laboratories, field and schools.
  • Option of quick, easy automatic calibration or customised, near-to-sample manual calibration
  • Auto-ranging for conductivity measurements for fast response and best resolution over wide measurement range
  • HOLD function and auto-power off
  • Self-diagnostic with message codes for easy troubleshooting
  • Electrode with built-in ATC designed for minimal air bubble entrapment during measurement
  • Built with shock-resistant ABS plastic