Sondes voor meten van opgeloste zuurstof, InLab® 605-ISM
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51344611 51344612 51344613
664-0041EA 1110 EUR
664-0041 664-0135 METT51344613
Sondes voor meten van opgeloste zuurstof, InLab® 605-ISM
Elektroden Elektroden voor opgeloste zuurstof
The InLab® 605-ISM DO sensor incorporates the newest technology in polarographic dissolved oxygen determination.
  • High accuracy and reproducibility
  • Membrane is protected by a steel mesh
  • Shaft is made out of extremely resistant glass fibre reinforced PPS (polyphenylene sulfide)
  • Withstand harsh conditions in factories, laboratories or out in the field
The InLab® 605-ISM sensor can be used in combination with a SevenExcellence multichannel benchtop meter or the SevenGo 68 dual channel portable meters.