Methylcyclopentane, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

66490-10ML 66490-50ML
66490-10MLEA 118 EUR
66490-10ML 66490-50ML
Methylcyclopentane, Supelco®
Methylcyclopentane
Formule: C₆H₁₂
Kookpunt: 71,8 °C (1013 hPa)
Smeltpunt: –142 °C
Dichtheid: 0,74 g/cm³ (20 °C)
Vlampunt: –17 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001382
CAS nummer: 96-37-7
EINECS: 202-503-2
UN: 2298
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3e3c51a409b5a8aa806b4f63abf84367e1dd8be&vid=93336b4a275aa301959c1aa44a582ee3caebfb16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR