Dissolved oxygen meter, handheld, Eutech DO 450
Leverancier: EUTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ECDOWP45003 ECDOWP45003K
665-0482EA 1470 EUR
665-0482 664-0231
Dissolved oxygen meter, handheld, Eutech DO 450
Elektrochemische meters Zuurstofmeters
This portable oxygen meter with optical probe measures dissolved oxygen and temperaure and is ideal for use in any environment. It features a durable waterproof and dustproof design with IP67 rating.
  • Large, backlit display with intuitive user-friendly icons
  • Grip-Clip™: Quickly and easily secures the electrode to a beaker or container
  • Calibration due alarm; manual barometric pressure and salinity correction
  • Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
  • Meets GLP standards: Built-in real-time clock stamps
More Product Information