VWR®, Silicone grease, TECHNICAL
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6674.0050EA 37.2 EUR
6674.0050
VWR®, Silicone grease, TECHNICAL
Smeermiddelen
VWR® Chemicals silicone grease is a valve lubricant and sealant, often times used as a vacuum grease. It has good resistance to most chemicals, a wide service temperature range, low vapor pressure, low volatility and exceptional water resistance.
  • Solubility: not soluble in H₂O , soluble in hydrocarbons
  • Temperature range: -50 to +200 °C
  • Penetration: 240 – 270 mm/10
This paste is especially intended for use in industrial an laboratory lubrication applications. It is used to lubricate taps and valves fitted to equipment working under vacuum. It is alsoMore Product Information
Order Now
Specificaties
Environmentally Preferable