Potassium hydrogen carbonate 99.5-101.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassium bicarbonate

6732-27EA 3230 EUR
6732-27
Potassium hydrogen carbonate 99.5-101.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
Potassium hydrogen carbonate
Formule: KHCO₃
Smeltpunt: 292 °C
Dichtheid: 2,17 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011402
CAS nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical 99.5 - 101.5 %
USP - Assay (KHCO₃) (dried basis) 99.5 - 101.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Loss on Drying ≤0.25 %
USP - Normal Carbonate ≤2.5 %
FCC - Assay (KHCO₃) (dried basis) 99.5 - 101.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Carbonate Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
Arsenic (As) ≤3 ppm
Solubility Passes Test
Appearance (white granules) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR