N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (by GC), LiChropur™ for GC derivatisation, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

69484-1ML 69484-5ML
69484-1MLEA 74 EUR
69484-1ML 69484-5ML
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) ≥90% (by GC), LiChropur™ for GC derivatisation, Supelco®
N-Methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA)

N-methyl-N-trimethylsilylheptafluorobutyramide (MSHFBA) has been used in the silylation reaction during GC analysis of samples containing 1-monoolein, 2-monoolein, 1,2-diolein, 1,3-diolein, ethyl oleate, oleic acid and triolein.

Formule: CF₃CF₂CF₂CON(CH₃)Si(CH₃)₃
MW: 299.26 g/mol
Kookpunt: 148 °C (literatuur)
Dichtheid: 1.254 g/mL (25 °C) (literatuur)
Vlampunt: 33 °C
MDL Nummer: MFCD00077822
CAS nummer: 53296-64-3
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5893fbdcbd45aece52d605a4faf060ce574b19a&vid=eacf81a3229667f578b181a1d23d67c02bbbe0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Colorless to Very Light Yellow
Appearance (Form) Liquid
Purity (GC area %) ≥ 90.0 %
Refractive index n20/D 1.351 - 1.355
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR