Kaliumhydroxide 12 mol/l (12 N) in aqueous solution

Leverancier: Hach
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potash caustic, Caustic potash

23032
705-0118EA 23.9 EUR
705-0118
Kaliumhydroxide 12 mol/l (12 N) in aqueous solution
Kaliumhydroxide
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 1320
Smeltpunt: 360
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 III
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR