Comparator kits, multiparameter for water testing, Comparator 2000+
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
413550 413570 413580
705-0361EA 3341.6 EUR
705-0361 705-0787 705-0362
Comparator kits, multiparameter for water testing, Comparator 2000+
Colorimeters Comparatorsets
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
  • Compact and portable
  • Simple to use yet uncompromising in terms of precision and reproducibility of results
Leveringsinformatie: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond®More Product Information