Test kits, photometric, phosphate by vanadate-molybdate method, LCK cuvette tests
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LCK049
705-3062EA 116 EUR
705-3062
Test kits, photometric, phosphate by vanadate-molybdate method, LCK cuvette tests
Milieutestkits
Cuvette tests for water testing applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Shelf life: 36 months from production date
Storage conditions: 15 to 25 °C
Informatie over toebehoren: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.
Order Now
Specificaties
Environmentally Preferable