Labo homogenisator, Masticator
Leverancier: IUL Instruments SA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
475 420 500 520 470 400
710-0103EA 2680 EUR
710-0103 710-0101 710-0104 710-0105 710-0102 710-0100
Labo homogenisator, Masticator
Homogenisatoren Laboratoriummixers
These Masticator laboratory blenders include two paddles which gently homogenise samples for microbiological testing. The blenders can be used with foodstuffs and drugs. The paddles provide pulsed movements to the contents of the sterile sample bag, efficiently homogenising the sample with the diluent. After a short period of time a solution, including all microorganisms to be analysed, is produced without any heat generation. The 'Panoramic' version features a glass door to view progress while the unit is in operation.
  • Geen enkel risico van verontreiniging van de monsters
More Product Information